The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004775.3 | NP_004766.2 | 4 - 5 | 767 | 65 |
XM_005258387.4 | XP_005258444.1 | 4 - 5 | 532 | 65 | |
XM_006722578.3 | XP_006722641.1 | 3 - 4 | 1621 | 65 | |
XM_006722579.3 | XP_006722642.1 | 5 - 6 | 480 | 65 | |
XM_017026088.1 | XP_016881577.1 | 3 - 4 | 1618 | 65 | |
XM_017026089.1 | XP_016881578.1 | 4 - 5 | 938 | 65 | |
GenBank mRNA | AB024742.2 | - | 4 - 5 | 476 | 65 |
AF038664.1 | - | 4 - 5 | 745 | 65 | |
AF069054.1 | - | 3 - 4 | 498 | 65 | |
AF097159.1 | - | 4 - 5 | 767 | 65 | |
AK314860.1 | - | 4 - 5 | 625 | 65 | |
BC069642.1 | - | 4 - 5 | 547 | 65 | |
BC074835.2 | - | 4 - 5 | 488 | 65 | |
BC074884.2 | - | 4 - 5 | 488 | 65 |