The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000139.4 | NP_000130.1 | 3 - 4 | 423 | 157 |
XM_005273846.4 | XP_005273903.1 | 4 - 5 | 738 | 157 | |
XM_011544850.2 | XP_011543152.1 | 4 - 5 | 356 | 157 | |
XM_017017362.1 | XP_016872851.1 | 4 - 5 | 738 | 157 | |
GenBank mRNA | AK304487.1 | - | 3 - 4 | 423 | 157 |
AK310142.1 | - | 3 - 4 | 423 | 157 | |
BC074800.2 | - | 3 - 4 | 352 | 157 | |
BC074843.2 | - | 3 - 4 | 352 | 157 | |
D10583.1 | - | 3 - 4 | 371 | 157 | |
DA972757.1 | - | 3 - 4 | 423 | 157 |