The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052854.3 | NP_443086.1 | 8 - 9 | 1500 | 103 |
GenBank mRNA | AB063321.1 | - | 8 - 9 | 1121 | 103 |
AK075538.1 | - | 3 - 4 | 271 | 103 | |
AK314072.1 | - | 8 - 9 | 1478 | 103 | |
BC014097.1 | - | 8 - 9 | 1143 | 103 | |
BC015781.1 | - | 8 - 9 | 1129 | 103 |