The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033714.2 | NP_001028886.1 | 1 - 2 | 197 | 105 |
XM_005253691.1 | XP_005253748.1 | 1 - 2 | 162 | 105 | |
GenBank mRNA | BC065257.1 | - | 1 - 2 | 139 | 105 |