Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPPED2
Entrez Gene ID
744
Gene Name
metallophosphoesterase domain containing 2
Gene Aliases
239FB, C11orf8
Chromosome Location
Chr.11: 30384493 - 30586713 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145399.1 NP_001138871.1 3 - 4 771 90
NM_001584.2 NP_001575.1 2 - 3 428 90
XM_005253110.3 XP_005253167.1 3 - 4 1101 90
XM_005253111.2 XP_005253168.1 3 - 4 738 90
XM_005253112.1 XP_005253169.1 3 - 4 566 90
XM_005253114.3 XP_005253171.1 3 - 4 1101 90
XM_017018231.1 XP_016873720.1 4 - 5 587 90
XM_017018232.1 XP_016873721.1 3 - 4 738 90
XM_017018233.1 XP_016873722.1 4 - 5 1124 90
XM_017018234.1 XP_016873723.1 4 - 5 1236 90
GenBank mRNA AB209163.1 - 3 - 4 669 90
AK314833.1 - 3 - 4 741 90
BC031582.1 - 2 - 3 444 90
U57911.1 - 2 - 3 428 90

View Full Product Details