The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320437.1 | NP_001307366.1 | 6 - 7 | 659 | 84 |
NM_001320441.1 | NP_001307370.1 | 7 - 8 | 1223 | 84 | |
NM_152586.3 | NP_689799.3 | 5 - 6 | 586 | 84 | |
NR_135250.1 | 4 - 5 | 455 | 84 | ||
XM_005269582.2 | XP_005269639.1 | 5 - 6 | 3835 | 84 | |
XM_017015773.1 | XP_016871262.1 | 5 - 6 | 3835 | 84 | |
XM_017015774.1 | XP_016871263.1 | 6 - 7 | 2827 | 84 | |
XM_017015775.1 | XP_016871264.1 | 7 - 8 | 2705 | 84 | |
XM_017015776.1 | XP_016871265.1 | 6 - 7 | 3210 | 84 | |
XM_017015777.1 | XP_016871266.1 | 6 - 7 | 2869 | 84 | |
XM_017015782.1 | XP_016871271.1 | 7 - 8 | 1607 | 84 | |
XM_017015783.1 | XP_016871272.1 | 7 - 8 | 2705 | 84 | |
XM_017015784.1 | XP_016871273.1 | 6 - 7 | 651 | 84 | |
XM_017015786.1 | XP_016871275.1 | 7 - 8 | 1268 | 84 | |
GenBank mRNA | AK127267.1 | - | 4 - 5 | 455 | 84 |
AK302598.1 | - | 7 - 8 | 1413 | 84 | |
BX649087.1 | - | 6 - 7 | 1130 | 84 | |
CN288586.1 | - | 5 - 6 | 622 | 84 |