The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024593.1 | NP_001019764.1 | 2 - 3 | 288 | 97 |
GenBank mRNA | AK096884.1 | - | 2 - 3 | 289 | 97 |
AK309870.1 | - | 3 - 4 | 344 | 97 | |
BC094693.1 | - | 2 - 3 | 267 | 97 | |
BC109255.1 | - | 1 - 2 | 201 | 97 | |
BC109256.1 | - | 1 - 2 | 201 | 97 | |
BX537678.1 | - | 2 - 3 | 339 | 97 | |
CR936824.1 | - | 2 - 3 | 270 | 97 |