Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
Chromosome Location
Chr.11: 85957171 - 86069881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008660.2 NP_001008660.1 3 - 4 660 141
NM_001206946.1 NP_001193875.1 3 - 4 660 141
NM_001206947.1 NP_001193876.1 3 - 4 375 141
NM_007166.3 NP_009097.2 3 - 4 660 141
XM_005274322.2 XP_005274379.1 3 - 4 644 141
XM_005274323.2 XP_005274380.1 3 - 4 644 141
XM_005274324.2 XP_005274381.1 3 - 4 644 141
XM_005274325.2 XP_005274382.1 3 - 4 644 141
XM_005274326.2 XP_005274383.1 3 - 4 644 141
XM_005274327.2 XP_005274384.1 3 - 4 644 141
XM_005274328.2 XP_005274385.1 3 - 4 644 141
XM_005274329.3 XP_005274386.1 3 - 4 644 141
XM_005274330.2 XP_005274387.1 3 - 4 644 141
XM_005274331.2 XP_005274388.1 3 - 4 644 141
XM_005274332.2 XP_005274389.1 3 - 4 644 141
XM_005274333.2 XP_005274390.1 3 - 4 644 141
XM_005274334.2 XP_005274391.1 3 - 4 644 141
XM_005274335.2 XP_005274392.1 3 - 4 644 141
XM_005274336.2 XP_005274393.1 3 - 4 644 141
XM_005274337.2 XP_005274394.1 3 - 4 644 141
XM_005274340.2 XP_005274397.1 3 - 4 644 141
XM_006718699.2 XP_006718762.1 3 - 4 644 141
XM_006718700.2 XP_006718763.1 3 - 4 644 141
XM_006718701.3 XP_006718764.1 3 - 4 644 141
XM_011545293.1 XP_011543595.1 3 - 4 644 141
XM_017018381.1 XP_016873870.1 3 - 4 644 141
XM_017018382.1 XP_016873871.1 3 - 4 644 141
XM_017018383.1 XP_016873872.1 3 - 4 644 141
XM_017018384.1 XP_016873873.1 3 - 4 644 141
XM_017018385.1 XP_016873874.1 3 - 4 644 141
XM_017018386.1 XP_016873875.1 3 - 4 644 141
XM_017018387.1 XP_016873876.1 3 - 4 644 141
XM_017018388.1 XP_016873877.1 3 - 4 644 141
GenBank mRNA AB210017.1 - 3 - 4 660 141
AB451308.1 - 3 - 4 343 141
AK291414.1 - 4 - 5 527 141
AK300275.1 - 3 - 4 352 141
BC048259.1 - 3 - 4 421 141
BC064357.1 - 3 - 4 622 141
BC073961.1 - 3 - 4 496 141
DC300074.1 - 3 - 4 375 141
U45976.1 - 3 - 4 490 141

View Full Product Details