The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153490.2 | NP_705694.2 | 6 - 7 | 1304 | 75 |
GenBank mRNA | AK092276.1 | - | 8 - 9 | 1234 | 75 |
AK223051.1 | - | 6 - 7 | 1303 | 75 | |
AK223077.1 | - | 6 - 7 | 1303 | 75 | |
AK290244.1 | - | 6 - 7 | 1303 | 75 | |
BC002661.2 | - | 6 - 7 | 1288 | 75 | |
BC077718.1 | - | 6 - 7 | 1281 | 75 | |
BC126184.1 | - | 6 - 7 | 1268 | 75 | |
BC136494.1 | - | 6 - 7 | 1268 | 75 | |
X14640.1 | - | 6 - 7 | 1283 | 75 |