The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173593.3 | NP_775864.3 | 18 - 19 | 2766 | 74 |
GenBank mRNA | AB089940.1 | - | 18 - 19 | 2766 | 74 |
AK097681.1 | - | 5 - 6 | 1151 | 74 | |
AK131277.1 | - | 5 - 6 | 2157 | 74 |