Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACO1
Entrez Gene ID
51204
Gene Name
translational activator of cytochrome c oxidase I
Gene Aliases
CCDC44
Chromosome Location
Chr.17: 63600872 - 63608365 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016360.3 NP_057444.2 1 - 2 494 85
GenBank mRNA AK315375.1 - 1 - 2 494 85
BC005049.1 - 1 - 2 482 85
BC007744.2 - 1 - 2 306 85

View Full Product Details