Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPHOSPH8
Entrez Gene ID
54737
Gene Name
M-phase phosphoprotein 8
Gene Aliases
HSMPP8, TWA3, mpp8
Chromosome Location
Chr.13: 19633647 - 19673459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017520.3 NP_059990.2 7 - 8 1892 124
XM_011535124.2 XP_011533426.1 7 - 8 1893 124
GenBank mRNA AJ293409.1 - 2 - 3 195 124
AK300258.1 - 7 - 8 1892 124
AL832864.1 - 7 - 8 1841 124
BC003542.1 - 7 - 8 1800 124

View Full Product Details