The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018159.1 | NP_001018169.1 | 16 - 17 | 1327 | 76 |
NM_001018160.1 | NP_001018170.1 | 14 - 15 | 1125 | 76 | |
NM_001286500.1 | NP_001273429.1 | 15 - 16 | 1238 | 76 | |
NM_003905.3 | NP_003896.1 | 15 - 16 | 1229 | 76 | |
XM_005256215.1 | XP_005256272.1 | 14 - 15 | 1199 | 76 | |
XM_011523422.2 | XP_011521724.1 | 13 - 14 | 1041 | 76 | |
GenBank mRNA | AK097680.1 | - | 15 - 16 | 1238 | 76 |
AK298159.1 | - | 16 - 17 | 1276 | 76 | |
AK312784.1 | - | 15 - 16 | 1229 | 76 | |
AL136798.1 | - | 15 - 16 | 1198 | 76 | |
AY197612.1 | - | 15 - 16 | 1167 | 76 | |
BC000480.2 | - | 15 - 16 | 1160 | 76 | |
BC013301.2 | - | 15 - 16 | 1160 | 76 | |
BC041323.2 | - | 7 - 8 | 557 | 76 | |
U50939.1 | - | 15 - 16 | 1225 | 76 |