The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018159.1 | NP_001018169.1 | 18 - 19 | 1498 | 116 |
NM_001018160.1 | NP_001018170.1 | 16 - 17 | 1296 | 116 | |
NM_001286500.1 | NP_001273429.1 | 17 - 18 | 1409 | 116 | |
NM_003905.3 | NP_003896.1 | 17 - 18 | 1400 | 116 | |
XM_005256215.1 | XP_005256272.1 | 16 - 17 | 1370 | 116 | |
XM_011523422.2 | XP_011521724.1 | 15 - 16 | 1212 | 116 | |
GenBank mRNA | AK097680.1 | - | 17 - 18 | 1409 | 116 |
AK298159.1 | - | 18 - 19 | 1447 | 116 | |
AK312784.1 | - | 17 - 18 | 1400 | 116 | |
AL136798.1 | - | 17 - 18 | 1369 | 116 | |
AY197612.1 | - | 17 - 18 | 1338 | 116 | |
BC000480.2 | - | 17 - 18 | 1331 | 116 | |
BC013301.2 | - | 17 - 18 | 1331 | 116 | |
BC041323.2 | - | 9 - 10 | 728 | 116 | |
U50939.1 | - | 17 - 18 | 1396 | 116 |