The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018159.1 | NP_001018169.1 | 7 - 8 | 577 | 123 |
NM_001018160.1 | NP_001018170.1 | 5 - 6 | 375 | 123 | |
NM_001286500.1 | NP_001273429.1 | 6 - 7 | 488 | 123 | |
NM_003905.3 | NP_003896.1 | 6 - 7 | 479 | 123 | |
XM_005256215.1 | XP_005256272.1 | 5 - 6 | 449 | 123 | |
XM_011523422.2 | XP_011521724.1 | 4 - 5 | 291 | 123 | |
GenBank mRNA | AK097680.1 | - | 6 - 7 | 488 | 123 |
AK298159.1 | - | 7 - 8 | 526 | 123 | |
AK307677.1 | - | 5 - 6 | 367 | 123 | |
AK312784.1 | - | 6 - 7 | 479 | 123 | |
AL136798.1 | - | 6 - 7 | 448 | 123 | |
AY197612.1 | - | 6 - 7 | 417 | 123 | |
BC000480.2 | - | 6 - 7 | 410 | 123 | |
BC013301.2 | - | 6 - 7 | 410 | 123 | |
BM045535.1 | - | 7 - 8 | 548 | 123 | |
U50939.1 | - | 6 - 7 | 475 | 123 |