Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFP90
Entrez Gene ID
146198
Gene Name
ZFP90 zinc finger protein
Gene Aliases
FIK, NK10, ZNF756, zfp-90
Chromosome Location
Chr.16: 68533484 - 68576072 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_130976.1 3 - 4 462 86
NR_130977.1 3 - 4 412 86
NR_130978.1 3 - 4 470 86
GenBank mRNA AK074332.1 - 3 - 4 378 86
BC040486.1 - 3 - 4 364 86

View Full Product Details