The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164356.1 | NP_001157828.1 | 1 - 2 | 90 | 63 |
GenBank mRNA | AF132970.1 | - | 1 - 2 | 90 | 63 |
AK225608.1 | - | 1 - 2 | 58 | 63 |