The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003955.4 | NP_003946.3 | 1 - 2 | 326 | 111 |
GenBank mRNA | AK309756.1 | - | 1 - 2 | 324 | 111 |
AK314212.1 | - | 1 - 2 | 324 | 111 | |
AU134017.1 | - | 1 - 2 | 324 | 111 | |
BC060858.1 | - | 1 - 2 | 324 | 111 | |
BE245649.1 | - | 1 - 2 | 145 | 111 | |
DA671124.1 | - | 1 - 2 | 326 | 111 |