The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141.2 | NP_001132.2 | 8 - 9 | 1321 | 105 |
GenBank mRNA | AF468051.1 | - | 8 - 9 | 1205 | 105 |
AF468054.1 | - | 8 - 9 | 1205 | 105 | |
AK292779.1 | - | 8 - 9 | 1317 | 105 | |
AK298095.1 | - | 8 - 9 | 1327 | 105 | |
AK300554.1 | - | 8 - 9 | 1305 | 105 | |
BC035217.1 | - | 8 - 9 | 1263 | 105 | |
BC063647.1 | - | 8 - 9 | 1277 | 105 | |
U78294.1 | - | 8 - 9 | 1276 | 105 |