Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEKT1
Entrez Gene ID
83659
Gene Name
tektin 1
Gene Aliases
-
Chromosome Location
Chr.17: 6799981 - 6831741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053285.1 NP_444515.1 2 - 3 299 112
XM_011524027.2 XP_011522329.1 2 - 3 295 112
GenBank mRNA AF357879.1 - 2 - 3 248 112
AK292818.1 - 2 - 3 264 112
BC014599.2 - 2 - 3 299 112

View Full Product Details