Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHRD
Entrez Gene ID
8646
Gene Name
chordin
Gene Aliases
-
Chromosome Location
Chr.3: 184380073 - 184389835 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304472.1 NP_001291401.1 5 - 6 860 61
NM_001304473.1 NP_001291402.1 6 - 7 915 61
NM_001304474.1 NP_001291403.1 5 - 6 860 61
NM_003741.3 NP_003732.2 5 - 6 860 61
NR_130747.1 5 - 6 860 61
XM_011513254.1 XP_011511556.1 5 - 6 713 61
XM_011513256.1 XP_011511558.1 4 - 5 501 61
XM_017007388.1 XP_016862877.1 5 - 6 713 61
XM_017007389.1 XP_016862878.1 5 - 6 713 61
XM_017007390.1 XP_016862879.1 5 - 6 713 61
XM_017007391.1 XP_016862880.1 5 - 6 713 61
XM_017007392.1 XP_016862881.1 5 - 6 713 61
XM_017007393.1 XP_016862882.1 5 - 6 713 61
XM_017007394.1 XP_016862883.1 4 - 5 501 61
GenBank mRNA AB208947.1 - 5 - 6 633 61
AF076612.1 - 3 - 4 272 61
AF209928.1 - 5 - 6 860 61
AF209929.1 - 6 - 7 911 61
AF209930.1 - 6 - 7 915 61
AF283325.1 - 5 - 6 860 61
AK074147.1 - 5 - 6 613 61
AK295632.1 - 5 - 6 500 61
AK296316.1 - 4 - 5 459 61
AY358926.1 - 5 - 6 750 61
BC112345.1 - 5 - 6 645 61

View Full Product Details