The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297573.1 | NP_001284502.1 | 13 - 14 | 3284 | 79 |
NM_033502.3 | NP_277037.1 | 13 - 14 | 3224 | 79 | |
XM_017011046.1 | XP_016866535.1 | 13 - 14 | 3702 | 79 | |
XM_017011048.1 | XP_016866537.1 | 13 - 14 | 3702 | 79 | |
XM_017011049.1 | XP_016866538.1 | 13 - 14 | 3642 | 79 | |
XM_017011052.1 | XP_016866541.1 | 12 - 13 | 3393 | 79 | |
XM_017011053.1 | XP_016866542.1 | 14 - 15 | 3569 | 79 | |
XM_017011054.1 | XP_016866543.1 | 12 - 13 | 3393 | 79 | |
GenBank mRNA | AF297872.1 | - | 13 - 14 | 3218 | 79 |
AJ277275.1 | - | 12 - 13 | 2759 | 79 | |
AJ277276.1 | - | 12 - 13 | 2759 | 79 | |
AK127493.1 | - | 9 - 10 | 2698 | 79 | |
AM404182.1 | - | 13 - 14 | 3284 | 79 | |
AM404183.1 | - | 13 - 14 | 3224 | 79 | |
AM404259.1 | - | 13 - 14 | 3224 | 79 |