Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP44
Entrez Gene ID
84101
Gene Name
ubiquitin specific peptidase 44
Gene Aliases
-
Chromosome Location
Chr.12: 95516560 - 95551529 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032147.4 NP_115523.2 1 - 2 225 153
XM_005269172.1 XP_005269229.1 1 - 2 219 153
XM_011538806.2 XP_011537108.1 1 - 2 219 153
XM_017020012.1 XP_016875501.1 1 - 2 219 153
GenBank mRNA AL136825.1 - 1 - 2 186 153
BI458616.1 - 1 - 2 231 152
DB443014.1 - 1 - 2 221 153
DB456039.1 - 1 - 2 178 153

View Full Product Details