The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287813.2 | NP_001274742.1 | 14 - 15 | 1279 | 120 |
NM_001287814.2 | NP_001274743.1 | 12 - 13 | 1431 | 120 | |
NM_001321773.1 | NP_001308702.1 | 12 - 13 | 1431 | 120 | |
NM_001321775.1 | NP_001308704.1 | 14 - 15 | 1279 | 120 | |
NM_001321776.1 | NP_001308705.1 | 13 - 14 | 1506 | 120 | |
NM_001321777.1 | NP_001308706.1 | 12 - 13 | 1431 | 120 | |
NM_001321778.1 | NP_001308707.1 | 13 - 14 | 1237 | 120 | |
NM_018967.4 | NP_061840.1 | 13 - 14 | 1506 | 120 | |
NR_135794.1 | 13 - 14 | 1486 | 120 | ||
NR_135795.1 | 13 - 14 | 1506 | 120 | ||
NR_135796.1 | 13 - 14 | 1506 | 120 | ||
NR_135798.1 | 12 - 13 | 1431 | 120 | ||
XM_011517548.2 | XP_011515850.1 | 12 - 13 | 1616 | 120 | |
XM_017013579.1 | XP_016869068.1 | 15 - 16 | 4435 | 120 | |
XM_017013580.1 | XP_016869069.1 | 13 - 14 | 1029 | 120 | |
GenBank mRNA | AJ003030.1 | - | 12 - 13 | 1067 | 120 |
AK314084.1 | - | 14 - 15 | 1216 | 120 | |
AL161971.1 | - | 13 - 14 | 1401 | 120 | |
BC075072.2 | - | 11 - 12 | 871 | 120 | |
BC104829.1 | - | 12 - 13 | 953 | 120 | |
BC143695.1 | - | 12 - 13 | 953 | 120 |