Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARS
Entrez Gene ID
833
Gene Name
cysteinyl-tRNA synthetase
Gene Aliases
CARS1, CYSRS, MGC:11246
Chromosome Location
Chr.11: 3000922 - 3057451 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014437.2 NP_001014437.1 6 - 7 736 95
NM_001194997.1 NP_001181926.1 6 - 7 736 95
NM_001751.5 NP_001742.1 5 - 6 487 95
NM_139273.3 NP_644802.1 5 - 6 487 95
NR_036542.1 6 - 7 783 95
XM_006718341.3 XP_006718404.1 5 - 6 510 95
XM_017018389.1 XP_016873878.1 5 - 6 510 95
XM_017018390.1 XP_016873879.1 6 - 7 750 95
XM_017018391.1 XP_016873880.1 3 - 4 734 95
XM_017018392.1 XP_016873881.1 5 - 6 672 95
GenBank mRNA AB208814.1 - 5 - 6 463 95
AF288206.1 - 5 - 6 420 95
AF288207.1 - 5 - 6 420 95
AK096313.1 - 6 - 7 750 95
AK222614.1 - 5 - 6 428 95
AK289432.1 - 5 - 6 460 95
AK295371.1 - 5 - 6 378 95
AK296763.1 - 6 - 7 601 95
AK298514.1 - 6 - 7 703 95
AK302644.1 - 6 - 7 703 95
AK308005.1 - 6 - 7 656 95
AU133264.1 - 5 - 6 477 95
BC002880.2 - 5 - 6 473 95
BT009913.1 - 5 - 6 403 95
BX647906.1 - 6 - 7 710 95
DC376478.1 - 5 - 6 487 95
L06845.1 - 3 - 4 274 95

View Full Product Details