Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH2
Entrez Gene ID
57158
Gene Name
junctophilin 2
Gene Aliases
CMH17, JP-2, JP2
Chromosome Location
Chr.20: 44111697 - 44187578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020433.4 NP_065166.2 2 - 3 2040 105
GenBank mRNA AK304863.1 - 2 - 3 685 105
BC043206.2 - 1 - 2 330 105

View Full Product Details