The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199573.1 | NP_001186502.1 | 2 - 3 | 805 | 91 |
NM_006074.4 | NP_006065.2 | 2 - 3 | 805 | 91 | |
GenBank mRNA | AK290434.1 | - | 2 - 3 | 535 | 91 |
AK298715.1 | - | 3 - 4 | 485 | 91 | |
BC022281.1 | - | 2 - 3 | 612 | 91 | |
BC035582.1 | - | 2 - 3 | 578 | 91 | |
HQ842635.1 | - | 1 - 2 | 426 | 91 | |
X82200.1 | - | 2 - 3 | 548 | 91 |