Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC7A5
Entrez Gene ID
8140
Gene Name
solute carrier family 7 member 5
Gene Aliases
4F2LC, CD98, D16S469E, E16, LAT1, MPE16, hLAT1
Chromosome Location
Chr.16: 87830022 - 87869499 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003486.5 NP_003477.4 5 - 6 1013 69
XM_006721286.3 XP_006721349.2 5 - 6 656 69
XM_017023735.1 XP_016879224.1 5 - 6 1018 69
XM_017023736.1 XP_016879225.1 4 - 5 892 69
XM_017023737.1 XP_016879226.1 5 - 6 605 69
GenBank mRNA AB017908.1 - 5 - 6 981 69
AB018009.1 - 5 - 6 1006 69
AB018542.1 - 5 - 6 956 69
AF077866.1 - 5 - 6 993 69
AF104032.1 - 5 - 6 1007 69
BC039692.2 - 5 - 6 1038 69
BC042600.1 - 5 - 6 1007 69
M80244.1 - 5 - 6 453 69

View Full Product Details