The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153333.2 | NP_699164.1 | 1 - 2 | 119 | 71 |
GenBank mRNA | AK026349.1 | - | 1 - 2 | 96 | 71 |
BC035573.1 | - | 1 - 2 | 136 | 71 | |
BU932815.1 | - | 1 - 2 | 131 | 71 |