Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EFHB
Entrez Gene ID
151651
Gene Name
EF-hand domain family member B
Gene Aliases
CFAP21
Chromosome Location
Chr.3: 19879472 - 19947036 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144715.3 NP_653316.3 2 - 3 1049 88
XM_005264889.1 XP_005264946.1 2 - 3 1049 88
XM_011533383.2 XP_011531685.2 2 - 3 157 88
XM_011533385.2 XP_011531687.1 3 - 4 425 88
XM_017005741.1 XP_016861230.1 4 - 5 609 88
XM_017005742.1 XP_016861231.1 2 - 3 1049 88
XM_017005743.1 XP_016861232.1 3 - 4 376 88
XM_017005745.1 XP_016861234.1 3 - 4 351 88
GenBank mRNA AK097644.1 - 2 - 3 1051 88
BC028198.1 - 2 - 3 1015 88

View Full Product Details