Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC27
Entrez Gene ID
148870
Gene Name
coiled-coil domain containing 27
Gene Aliases
-
Chromosome Location
Chr.1: 3752398 - 3771645 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152492.2 NP_689705.2 4 - 5 796 64
XM_011540780.1 XP_011539082.1 4 - 5 799 64
XM_017000402.1 XP_016855891.1 4 - 5 799 64
XM_017000403.1 XP_016855892.1 4 - 5 799 64
XM_017000404.1 XP_016855893.1 4 - 5 799 64
XM_017000405.1 XP_016855894.1 4 - 5 799 64
XM_017000406.1 XP_016855895.1 5 - 6 613 64
GenBank mRNA AK057387.1 - 4 - 5 799 64
BC112108.1 - 4 - 5 797 64
BC112112.1 - 4 - 5 796 64

View Full Product Details