The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000212.2 | NP_000203.2 | 14 - 15 | 2328 | 59 |
GenBank mRNA | BC127666.1 | - | 14 - 15 | 2328 | 59 |
BC127667.1 | - | 14 - 15 | 2332 | 59 | |
J02703.1 | - | 14 - 15 | 2328 | 59 | |
KC120777.1 | - | 14 - 15 | 2329 | 59 | |
KC120778.1 | - | 14 - 15 | 2328 | 59 | |
M20311.1 | - | 14 - 15 | 2324 | 59 | |
M35999.1 | - | 14 - 15 | 2326 | 59 |