The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000212.2 | NP_000203.2 | 1 - 2 | 97 | 135 |
GenBank mRNA | BC127666.1 | - | 1 - 2 | 97 | 135 |
BC127667.1 | - | 1 - 2 | 101 | 135 | |
J02703.1 | - | 1 - 2 | 97 | 135 | |
KC120777.1 | - | 1 - 2 | 97 | 135 | |
KC120778.1 | - | 1 - 2 | 97 | 135 | |
M20311.1 | - | 1 - 2 | 93 | 135 | |
M35999.1 | - | 1 - 2 | 95 | 135 |