The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000212.2 | NP_000203.2 | 5 - 6 | 793 | 74 |
GenBank mRNA | AY826979.1 | - | 5 - 6 | 782 | 74 |
BC127666.1 | - | 5 - 6 | 793 | 74 | |
BC127667.1 | - | 5 - 6 | 797 | 74 | |
J02703.1 | - | 5 - 6 | 793 | 74 | |
KC120777.1 | - | 5 - 6 | 793 | 74 | |
KC120778.1 | - | 5 - 6 | 793 | 74 | |
M20311.1 | - | 5 - 6 | 789 | 74 | |
M35999.1 | - | 5 - 6 | 791 | 74 | |
U95204.1 | - | 5 - 6 | 773 | 74 |