The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001323.1 | NP_001001323.1 | 3 - 4 | 840 | 79 |
NM_001682.2 | NP_001673.2 | 3 - 4 | 840 | 79 | |
XM_011538407.2 | XP_011536709.1 | 4 - 5 | 1015 | 79 | |
XM_017019357.1 | XP_016874846.1 | 3 - 4 | 792 | 79 | |
XM_017019358.1 | XP_016874847.1 | 3 - 4 | 792 | 79 | |
XM_017019359.1 | XP_016874848.1 | 3 - 4 | 792 | 79 | |
XM_017019360.1 | XP_016874849.1 | 3 - 4 | 792 | 79 | |
GenBank mRNA | BC015346.1 | - | 3 - 4 | 883 | 79 |
J04027.1 | - | 3 - 4 | 840 | 79 | |
M95541.1 | - | 3 - 4 | 840 | 79 | |
M95542.1 | - | - | 79 |