Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ORAOV1
Entrez Gene ID
220064
Gene Name
oral cancer overexpressed 1
Gene Aliases
TAOS1
Chromosome Location
Chr.11: 69661350 - 69675397 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006718470.2 XP_006718533.1 5 - 6 571 120

View Full Product Details