The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305204.1 | NP_001292133.1 | 1 - 2 | 257 | 142 |
NM_001305208.1 | NP_001292137.1 | 1 - 2 | 257 | 142 | |
NM_133458.3 | NP_597715.2 | 1 - 2 | 257 | 142 | |
GenBank mRNA | AK304152.1 | - | 1 - 2 | 159 | 142 |
BC140886.1 | - | 1 - 2 | 257 | 142 | |
BC140887.1 | - | 1 - 2 | 257 | 142 | |
DA803284.1 | - | 1 - 2 | 181 | 142 |