The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305203.1 | NP_001292132.1 | 3 - 4 | 457 | 70 |
NM_001305204.1 | NP_001292133.1 | 3 - 4 | 447 | 70 | |
NM_001305206.1 | NP_001292135.1 | 3 - 4 | 457 | 70 | |
NM_001305207.1 | NP_001292136.1 | 3 - 4 | 457 | 70 | |
NM_001305208.1 | NP_001292137.1 | 3 - 4 | 447 | 70 | |
NM_133458.3 | NP_597715.2 | 3 - 4 | 447 | 70 | |
XM_005255804.2 | XP_005255861.1 | 3 - 4 | 387 | 70 | |
XM_017022952.1 | XP_016878441.1 | 2 - 3 | 494 | 70 | |
GenBank mRNA | AK304152.1 | - | 3 - 4 | 349 | 70 |
AK316251.1 | - | 3 - 4 | 374 | 70 | |
BC140886.1 | - | 3 - 4 | 447 | 70 | |
BC140887.1 | - | 3 - 4 | 447 | 70 | |
DA803284.1 | - | 3 - 4 | 371 | 70 | |
KC249978.1 | - | 3 - 4 | 344 | 70 |