The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005862.2 | NP_001005862.1 | 6 - 7 | 923 | 64 |
NM_001289936.1 | NP_001276865.1 | 7 - 8 | 974 | 64 | |
NM_001289937.1 | NP_001276866.1 | 3 - 4 | 698 | 64 | |
NM_001289938.1 | NP_001276867.1 | 6 - 7 | 923 | 64 | |
NM_004448.3 | NP_004439.2 | 3 - 4 | 698 | 64 | |
NR_110535.1 | 3 - 4 | 761 | 64 | ||
GenBank mRNA | AF177761.2 | - | 3 - 4 | 449 | 64 |
AK131568.1 | - | 1 - 2 | 1129 | 64 | |
AK295195.1 | - | 7 - 8 | 974 | 64 | |
AK313683.1 | - | 3 - 4 | 652 | 64 | |
BC080193.1 | - | 3 - 4 | 659 | 64 | |
BC110392.1 | - | 6 - 7 | 888 | 64 | |
BC167147.1 | - | 3 - 4 | 440 | 64 | |
KJ534831.1 | - | 3 - 4 | 437 | 64 | |
KJ534964.1 | - | 3 - 4 | 437 | 64 | |
M11730.1 | - | 3 - 4 | 587 | 64 | |
X03363.1 | - | 3 - 4 | 611 | 64 |