The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005862.2 | NP_001005862.1 | 10 - 11 | 1388 | 77 |
NM_001289936.1 | NP_001276865.1 | 11 - 12 | 1439 | 77 | |
NM_001289937.1 | NP_001276866.1 | 7 - 8 | 1163 | 77 | |
NM_001289938.1 | NP_001276867.1 | 10 - 11 | 1388 | 77 | |
NM_004448.3 | NP_004439.2 | 7 - 8 | 1163 | 77 | |
NR_110535.1 | 7 - 8 | 1226 | 77 | ||
GenBank mRNA | AF177761.2 | - | 7 - 8 | 914 | 77 |
AK131568.1 | - | 5 - 6 | 1594 | 77 | |
AK295195.1 | - | 11 - 12 | 1439 | 77 | |
AK296908.1 | - | 7 - 8 | 978 | 77 | |
AK313683.1 | - | 7 - 8 | 1117 | 77 | |
BC080193.1 | - | 7 - 8 | 1124 | 77 | |
BC110392.1 | - | 10 - 11 | 1353 | 77 | |
BC167147.1 | - | 7 - 8 | 905 | 77 | |
KJ534831.1 | - | 7 - 8 | 902 | 77 | |
KJ534964.1 | - | 7 - 8 | 902 | 77 | |
M11730.1 | - | 7 - 8 | 1052 | 77 | |
X03363.1 | - | 7 - 8 | 1076 | 77 |