The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000458.3 | NP_000449.1 | 8 - 9 | 1852 | 70 |
NM_001165923.3 | NP_001159395.1 | 8 - 9 | 1774 | 70 | |
XM_011525161.1 | XP_011523463.1 | 7 - 8 | 1638 | 70 | |
GenBank mRNA | AK290131.1 | - | 8 - 9 | 1833 | 70 |
AK296633.1 | - | 8 - 9 | 1755 | 70 | |
AK298393.1 | - | 9 - 10 | 1702 | 70 | |
BC017714.1 | - | 8 - 9 | 1846 | 70 | |
HM116553.1 | - | 8 - 9 | 1714 | 70 | |
HM116554.1 | - | 7 - 8 | 1500 | 70 | |
X58840.1 | - | 8 - 9 | 1852 | 70 |