Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCM1
Entrez Gene ID
5108
Gene Name
pericentriolar material 1
Gene Aliases
PTC4, RET/PCM-1
Chromosome Location
Chr.8: 17922763 - 18029948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001315507.1 NP_001302436.1 32 - 33 5625 153
NM_001315508.1 NP_001302437.1 31 - 32 5487 153
NM_006197.3 NP_006188.3 32 - 33 5649 153
XM_006716336.3 XP_006716399.1 35 - 36 6100 153
XM_006716340.3 XP_006716403.1 35 - 36 6088 153
XM_006716349.3 XP_006716412.1 31 - 32 5560 153
XM_011544528.2 XP_011542830.1 35 - 36 6121 153
XM_011544529.2 XP_011542831.1 37 - 38 6427 153
XM_011544530.2 XP_011542832.1 36 - 37 6243 153
XM_011544532.2 XP_011542834.1 35 - 36 6118 153
XM_011544533.2 XP_011542835.1 35 - 36 6109 153
XM_011544534.2 XP_011542836.1 35 - 36 6097 153
XM_011544535.2 XP_011542837.1 35 - 36 6085 153
XM_011544540.2 XP_011542842.1 35 - 36 6049 153
XM_017013472.1 XP_016868961.1 36 - 37 6237 153
XM_017013473.1 XP_016868962.1 36 - 37 6233 153
XM_017013474.1 XP_016868963.1 33 - 34 5851 153
XM_017013475.1 XP_016868964.1 35 - 36 5975 153
XM_017013476.1 XP_016868965.1 32 - 33 5559 153
XM_017013477.1 XP_016868966.1 35 - 36 6140 153
XM_017013478.1 XP_016868967.1 35 - 36 6118 153
XM_017013479.1 XP_016868968.1 35 - 36 6119 153
XM_017013480.1 XP_016868969.1 33 - 34 5791 153
XM_017013481.1 XP_016868970.1 35 - 36 6094 153
XM_017013482.1 XP_016868971.1 32 - 33 5755 153
XM_017013483.1 XP_016868972.1 34 - 35 6024 153
XM_017013484.1 XP_016868973.1 31 - 32 5450 153
XM_017013485.1 XP_016868974.1 34 - 35 6004 153
XM_017013486.1 XP_016868975.1 32 - 33 5686 153
XM_017013487.1 XP_016868976.1 32 - 33 5677 153
XM_017013488.1 XP_016868977.1 32 - 33 5674 153
XM_017013489.1 XP_016868978.1 34 - 35 5977 153
XM_017013490.1 XP_016868979.1 34 - 35 5953 153
XM_017013491.1 XP_016868980.1 31 - 32 5572 153
XM_017013492.1 XP_016868981.1 31 - 32 5569 153
XM_017013493.1 XP_016868982.1 33 - 34 5872 153
XM_017013494.1 XP_016868983.1 31 - 32 5557 153
XM_017013495.1 XP_016868984.1 33 - 34 5697 153
XM_017013496.1 XP_016868985.1 30 - 31 5280 153
XM_017013497.1 XP_016868986.1 31 - 32 5545 153
XM_017013498.1 XP_016868987.1 31 - 32 5536 153
XM_017013499.1 XP_016868988.1 33 - 34 5676 153
XM_017013500.1 XP_016868989.1 31 - 32 5533 153
XM_017013501.1 XP_016868990.1 33 - 34 5835 153
XM_017013502.1 XP_016868991.1 30 - 31 5256 153
GenBank mRNA AK091406.1 - 9 - 10 1334 153
AK295856.1 - 10 - 11 1635 153
AK301727.1 - 9 - 10 1141 153
AK302378.1 - 11 - 12 1446 153
AX785390.1 - 30 - 31 5227 153
BC133052.1 - 30 - 31 5137 153
BC140946.1 - 32 - 33 5454 153
BG898673.1 - 2 - 3 212 153
L27841.1 - 32 - 33 5636 153

View Full Product Details