The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284400.2 | NP_001271329.1 | 2 - 3 | 411 | 84 |
NM_001321675.1 | NP_001308604.1 | 2 - 3 | 407 | 84 | |
NM_001321676.1 | NP_001308605.1 | 2 - 3 | 411 | 84 | |
NM_001321677.1 | NP_001308606.1 | 2 - 3 | 411 | 84 | |
NM_144598.4 | NP_653199.2 | 2 - 3 | 411 | 84 | |
NR_135753.1 | 2 - 3 | 411 | 84 | ||
NR_135757.1 | 2 - 3 | 411 | 84 | ||
XM_006720389.3 | XP_006720452.1 | 2 - 3 | 411 | 84 | |
XM_011521218.2 | XP_011519520.1 | 2 - 3 | 447 | 84 | |
XM_011521220.1 | XP_011519522.1 | 2 - 3 | 411 | 84 | |
XM_011521221.2 | XP_011519523.1 | 2 - 3 | 411 | 84 | |
XM_017021914.1 | XP_016877403.1 | 2 - 3 | 695 | 84 | |
GenBank mRNA | AK292887.1 | - | 2 - 3 | 268 | 84 |
AK293832.1 | - | 2 - 3 | 268 | 84 | |
AK295161.1 | - | 2 - 3 | 286 | 84 | |
AK296352.1 | - | 2 - 3 | 313 | 84 | |
AY358082.1 | - | 2 - 3 | 268 | 84 | |
BC019704.1 | - | 2 - 3 | 447 | 84 | |
BC047306.1 | - | 2 - 3 | 326 | 84 |