The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003722.1 | NP_001003722.1 | 11 - 12 | 1755 | 79 |
NM_001499.2 | NP_001490.1 | 11 - 12 | 1755 | 79 | |
XM_006717059.2 | XP_006717122.1 | 13 - 14 | 1894 | 79 | |
XM_006717060.2 | XP_006717123.1 | 12 - 13 | 1867 | 79 | |
XM_011518549.1 | XP_011516851.1 | 13 - 14 | 1931 | 79 | |
XM_011518550.1 | XP_011516852.1 | 13 - 14 | 1782 | 79 | |
XM_011518551.1 | XP_011516853.1 | 12 - 13 | 1782 | 79 | |
GenBank mRNA | AF058922.1 | - | 11 - 12 | 1728 | 79 |
AK001767.1 | - | 6 - 7 | 1005 | 79 | |
AK074801.1 | - | 10 - 11 | 1511 | 79 | |
AK222854.1 | - | 11 - 12 | 1729 | 79 | |
AK290533.1 | - | 10 - 11 | 1492 | 79 | |
AL117584.1 | - | 9 - 10 | 1238 | 79 | |
BC030012.1 | - | 11 - 12 | 1748 | 79 |