Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLE1
Entrez Gene ID
2733
Gene Name
GLE1, RNA export mediator
Gene Aliases
GLE1L, LCCS, LCCS1, hGLE1
Chromosome Location
Chr.9: 128504692 - 128542301 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003722.1 NP_001003722.1 5 - 6 759 84
NM_001499.2 NP_001490.1 5 - 6 759 84
XM_006717059.2 XP_006717122.1 6 - 7 871 84
XM_006717060.2 XP_006717123.1 6 - 7 871 84
XM_011518549.1 XP_011516851.1 6 - 7 908 84
XM_011518550.1 XP_011516852.1 6 - 7 759 84
XM_011518551.1 XP_011516853.1 5 - 6 759 84
GenBank mRNA AF058922.1 - 5 - 6 732 84
AK074801.1 - 4 - 5 515 84
AK222842.1 - 5 - 6 705 84
AK222854.1 - 5 - 6 733 84
AK290533.1 - 4 - 5 496 84
AL117584.1 - 3 - 4 242 84
BC030012.1 - 5 - 6 752 84

View Full Product Details