The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003722.1 | NP_001003722.1 | 3 - 4 | 549 | 70 |
NM_001499.2 | NP_001490.1 | 3 - 4 | 549 | 70 | |
XM_006717059.2 | XP_006717122.1 | 4 - 5 | 661 | 70 | |
XM_006717060.2 | XP_006717123.1 | 4 - 5 | 661 | 70 | |
XM_011518549.1 | XP_011516851.1 | 4 - 5 | 698 | 70 | |
XM_011518550.1 | XP_011516852.1 | 4 - 5 | 549 | 70 | |
XM_011518551.1 | XP_011516853.1 | 3 - 4 | 549 | 70 | |
GenBank mRNA | AF058922.1 | - | 3 - 4 | 522 | 70 |
AK222842.1 | - | 3 - 4 | 495 | 70 | |
AK222854.1 | - | 3 - 4 | 523 | 70 | |
BC030012.1 | - | 3 - 4 | 542 | 70 | |
BM786801.1 | - | 3 - 4 | 549 | 70 |