The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111112.1 | NP_001104582.1 | 1 - 2 | 346 | 122 |
NM_001312646.1 | NP_001299575.1 | 1 - 2 | 346 | 122 | |
NM_005339.4 | NP_005330.1 | 1 - 2 | 346 | 122 | |
GenBank mRNA | AB022435.1 | - | 1 - 2 | 148 | 122 |
AK295846.1 | - | 1 - 2 | 281 | 122 | |
AK315524.1 | - | 1 - 2 | 272 | 122 | |
BC022804.1 | - | 1 - 2 | 222 | 122 | |
BC050600.1 | - | 1 - 2 | 200 | 122 | |
CX786829.1 | - | 1 - 2 | 185 | 122 | |
DA526283.1 | - | 1 - 2 | 346 | 122 | |
U58522.1 | - | 1 - 2 | 65 | 122 |