The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105248.1 | NP_001098718.1 | 17 - 18 | 3316 | 140 |
NM_001261841.1 | NP_001248770.1 | 17 - 18 | 3271 | 140 | |
NM_001308161.1 | NP_001295090.1 | 16 - 17 | 2953 | 140 | |
NM_024780.4 | NP_079056.2 | 13 - 14 | 2086 | 140 | |
GenBank mRNA | AK023655.1 | - | 6 - 7 | 569 | 140 |
AK093944.1 | - | 10 - 11 | 1143 | 140 | |
AK097718.1 | - | 13 - 14 | 2051 | 140 | |
AK131202.1 | - | 14 - 15 | 1806 | 140 | |
AK299055.1 | - | 16 - 17 | 2953 | 140 | |
AK304431.1 | - | 17 - 18 | 3271 | 140 | |
AY236494.1 | - | 13 - 14 | 2086 | 140 | |
AY263164.1 | - | 13 - 14 | 1829 | 140 | |
BC027602.1 | - | 13 - 14 | 2041 | 140 | |
BC038118.2 | - | 13 - 14 | 1781 | 140 | |
CR749359.1 | - | 15 - 16 | 1971 | 140 |