The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167621.1 | NP_001161093.1 | 9 - 10 | 1310 | 78 |
NM_013236.3 | NP_037368.1 | 10 - 11 | 1502 | 78 | |
GenBank mRNA | AF119662.1 | - | 10 - 11 | 1442 | 78 |
AK025734.1 | - | 10 - 11 | 1433 | 78 | |
AK095309.1 | - | 10 - 11 | 1270 | 78 | |
AK294348.1 | - | 9 - 10 | 1303 | 78 | |
AL050282.1 | - | 10 - 11 | 1434 | 78 | |
AL137374.1 | - | 1 - 2 | 71 | 78 | |
BC007508.2 | - | 10 - 11 | 1424 | 78 | |
CR456568.1 | - | 10 - 11 | 1287 | 78 | |
CR457381.1 | - | 10 - 11 | 1236 | 78 |