The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278167.1 | NP_001265096.1 | 2 - 3 | 471 | 89 |
NM_001278168.1 | NP_001265097.1 | 2 - 3 | 471 | 89 | |
NM_001278169.1 | NP_001265098.1 | 2 - 3 | 239 | 89 | |
NM_016082.4 | NP_057166.4 | 2 - 3 | 323 | 89 | |
NM_016408.3 | NP_057492.2 | 2 - 3 | 471 | 89 | |
XM_017027876.1 | XP_016883365.1 | 2 - 3 | 432 | 89 | |
GenBank mRNA | AF152097.1 | - | 2 - 3 | 410 | 89 |
AF161516.1 | - | 2 - 3 | 422 | 89 | |
AK023992.1 | - | 2 - 3 | 421 | 89 | |
AK027449.1 | - | 2 - 3 | 271 | 89 | |
AK222745.1 | - | 2 - 3 | 421 | 89 | |
AK292075.1 | - | 2 - 3 | 189 | 89 | |
AK295342.1 | - | 2 - 3 | 407 | 89 | |
AK309906.1 | - | 2 - 3 | 421 | 89 | |
AK314532.1 | - | 2 - 3 | 421 | 89 | |
AY462283.1 | - | 2 - 3 | 422 | 89 | |
AY462284.1 | - | 2 - 3 | 422 | 89 | |
BC001215.1 | - | 2 - 3 | 400 | 89 | |
BC050706.1 | - | 2 - 3 | 414 | 89 | |
BM834342.1 | - | 2 - 3 | 274 | 89 | |
BQ072829.1 | - | - | 433 | 89 | |
DA028522.1 | - | 2 - 3 | 421 | 89 | |
DA565688.1 | - | 2 - 3 | 273 | 89 | |
DC427896.1 | - | 2 - 3 | 471 | 89 | |
DR156307.1 | - | 1 - 2 | 80 | 89 | |
KU178644.1 | - | 1 - 2 | 309 | 89 | |
KU178645.1 | - | 1 - 2 | 309 | 89 |